1,3,6,8-Tetramethyl-9H-carbazole ≥98.0% (by GC, total nitrogen)

Leverantör: TCI
Varning

T3882-1G T3882-200MG
TCIAT3882-1GEA 7350 SEK
TCIAT3882-1G TCIAT3882-200MG
1,3,6,8-Tetramethyl-9H-carbazole ≥98.0% (by GC, total nitrogen)
1,3,6,8-Tetramethyl-9H-carbazole
Formel: C₁₆H₁₇N
MW: 223.32 g/mol
MDL Number: MFCD18450169
CAS nummer: 6558-85-6

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR