2-(Trimethylsilylethynyl)pyridine ≥98.0%
Leverantör: TCI
Varning
T3879-1G T3879-5G
TCIAT3879-1GEA 415 SEK
TCIAT3879-1G TCIAT3879-5G
2-(Trimethylsilylethynyl)pyridine ≥98.0%
2-(Trimethylsilylethynyl)pyridine
Formel: C₁₀H₁₃NSi
Kokpunkt: 90°C
Storage Temperature: Frys
MDL Number: MFCD00066349
CAS nummer: 86521-05-3
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %