2-([1,1':3',1''-Terphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Synonyms:
4,4,5,5-Tetramethyl-2-[1,1':3',1''-terphenyl]-4-yl-1,3,2-dioxaborolane, (1,1':3',1''-Terphenyl)-4-ylboronic acid Pinacol ester
T3873-1G
T3873-200MG
TCIAT3873-1GEA
1850
SEK
TCIAT3873-1G
TCIAT3873-200MG
2-([1,1':3',1''-Terphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-([1,1':3',1''-Terphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel:
C₂₄H₂₅BO₂ MW: 356.27 g/mol |
CAS nummer:
1385826-84-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...