2,4,6-Tris(4-butoxy-2-hydroxyphenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE

Synonyms: 6,6',6''-(1,3,5-Triazine-2,4,6-triyl)tris(3-butoxyphenol)

T3870-1G T3870-5G
TCIAT3870-1GEA 1050 SEK
TCIAT3870-1G TCIAT3870-5G
2,4,6-Tris(4-butoxy-2-hydroxyphenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4,6-Tris(4-butoxy-2-hydroxyphenyl)-1,3,5-triazine
Formel: C₃₃H₃₉N₃O₆
MW: 573,69 g/mol
CAS nummer: 3135-19-1

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR