Tolcapone ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
3,4-Dihydroxy-4'-methyl-5-nitrobenzophenone
T3856-200MG
T3856-50MG
TCIAT3856-200MGEA
4175
SEK
TCIAT3856-200MG
TCIAT3856-50MG
Tolcapone ≥98.0%
Tolcapone
Formel:
C₁₄H₁₁NO₅ MW: 273,24 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00866569 CAS nummer: 134308-13-7 Merck Index: 14,09508 |
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 142.0 to 146.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...