TTNPB ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Arotinoid acid, (E)-4-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1-propenyl]benzoic acid, (E)-4-(2-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)prop-1-en-1-yl)benzoic acid, TTNPB (Arotinoid Acid)

T3842-25MG T3842-5MG
TCIAT3842-25MGEA 3450 SEK
TCIAT3842-25MG TCIAT3842-5MG
TTNPB ≥98.0%
TTNPB
Formel: C₂₄H₂₈O₂
MW: 348,5 g/mol
Storage Temperature: Frys
MDL Number: MFCD00673917
CAS nummer: 71441-28-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 240.0 to 244.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR