1,1,5,5-Tetramethyl-3,3-diphenyltrisiloxane ≥98.0%

Leverantör: TCI
Varning

Synonyms: Bis(dimethylsiloxy)diphenylsilane

T3832-25G T3832-100G
TCIAT3832-25GEA 725 SEK
TCIAT3832-25G TCIAT3832-100G
1,1,5,5-Tetramethyl-3,3-diphenyltrisiloxane ≥98.0%
1,1,5,5-Tetramethyl-3,3-diphenyltrisiloxane
Formel: C₁₆H₂₄O₂Si₃
MW: 332.62 g/mol
Kokpunkt: 83°C
MDL Number: MFCD00411646
CAS nummer: 17875-55-7
EINECS: 241-828-4

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR