N-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acrylamide ≥98.0%

Leverantör: TCI

Synonyms: N-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylamide, 2-(3-Acrylamidophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Acrylamidophenylboronic acid Pinacol ester

T3826-1G T3826-5G
TCIAT3826-1GEA 935 SEK
TCIAT3826-1G TCIAT3826-5G
N-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acrylamide ≥98.0%
N-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acrylamide
Formel: C₁₅H₂₀BNO₃
MDL Number: MFCD22493996
CAS nummer: 874363-18-5

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 158.0 to 162.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR