N,N,N-Trimethyl-1-adamantylammonium hydroxide 25% i vatten
Leverantör: TCI
|
Fara
|
Synonyms:
N,N,N-Trimethyladamantan-1-aminium hydroxide
T3821-25G
T3821-100G
TCIAT3821-25GEA
910
SEK
TCIAT3821-25G
TCIAT3821-100G
N,N,N-Trimethyl-1-adamantylammonium hydroxide 25% i vatten
N,N,N-Trimethyl-1-adamantylammonium hydroxide
Formel:
C₁₃H₂₅NO MW: 211.35 g/mol |
MDL Number:
MFCD27920795 CAS nummer: 53075-09-5 UN: 3267 ADR: 8 II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Nonaqueous Titration) | 25.0 to 28.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...