[(3-Triethoxysilyl)propyl]succinic anhydride ≥93.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-(3-(Triethoxysilyl)propyl)dihydrofuran-2,5-dione

T3817-25G T3817-5G
TCIAT3817-25GEA 885 SEK
TCIAT3817-25G TCIAT3817-5G
[(3-Triethoxysilyl)propyl]succinic anhydride ≥93.0%
[(3-Triethoxysilyl)propyl]succinic anhydride
Formel: C₁₃H₂₄O₆Si
MW: 304,41 g/mol
MDL Number: MFCD01320976
CAS nummer: 93642-68-3
EINECS: 297-566-6

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 93.0 %
Purity (Morpholine Method) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR