Tetrakis(2-ethylhexyl) Orthosilicate ≥97.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Tetrakis(2-ethylhexyloxy)silane
T3748-25ML
T3748-5ML
TCIAT3748-25MLEA
1325
SEK
TCIAT3748-25ML
TCIAT3748-5ML
Tetrakis(2-ethylhexyl) Orthosilicate ≥97.0%
Tetrakis(2-ethylhexyl) Orthosilicate
Formel:
C₃₂H₆₈O₄Si MW: 544.98 g/mol Kokpunkt: 365°C Flampunkt: 199°C |
MDL Number:
MFCD00015263 CAS nummer: 115-82-2 EINECS: 204-109-6 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...