3,3',5,5'-Tetraiodothyroacetic acid ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Fara
Synonyms: 4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenylacetic acid, 4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzeneacetic acid
T3730-100MG T3730-500MG
TCIAT3730-100MGEA 1850 SEK
TCIAT3730-100MG TCIAT3730-500MG
3,3',5,5'-Tetraiodothyroacetic acid ≥98.0% (by HPLC, total nitrogen)
3,3',5,5'-Tetraiodothyroacetic acid
Formel: C₁₄H₈I₄O₄
MW: 747,83 g/mol
Smältpunkt: 230°C
Storage Temperature: Frys
MDL Number: MFCD00055932
CAS nummer: 67-30-1
EINECS: 200-649-1
UN: 2811
ADR: 6.1,I
Order Now
Specifikation testresultat
Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %