Tectoridin ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
7-(beta-D-Glucopyranosyloxy)-5-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 5-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Tectorigenin 7-O-beta-D-Glucopyranoside
T3724-10MG
T3724-50MG
TCIAT3724-10MGEA
765
SEK
TCIAT3724-10MG
TCIAT3724-50MG
Tectoridin ≥98.0%
Tectoridin
Formel:
C₂₂H₂₂O₁₁ MW: 462,41 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD01662715 CAS nummer: 611-40-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...