1-(Trifluoromethyl)cyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
T3717-200MG T3717-1G
TCIAT3717-200MGEA 655 SEK
TCIAT3717-200MG TCIAT3717-1G
1-(Trifluoromethyl)cyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(Trifluoromethyl)cyclobutanecarboxylic acid
Formel: C₆H₇F₃O₂
Kokpunkt: 76°C
Flampunkt: 96°C
Storage Temperature: Kylskåp
MDL Number: MFCD03425197
CAS nummer: 277756-45-3
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %