3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-(3-Formylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Formylphenylboronic acid, pinacol ester, 3-Formylphenylboronic acid Pinacol ester

T3709-5G T3709-25G
TCIAT3709-5GEA 885 SEK
TCIAT3709-5G TCIAT3709-25G
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥98.0%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Formel: C₁₃H₁₇BO₃
MW: 232,09 g/mol
MDL Number: MFCD05663827
CAS nummer: 380151-86-0

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR