Tetravinyltin ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Tetravinylstannane, Tetravinyl tin
T3703-25G
T3703-5G
TCIAT3703-25GEA
3250
SEK
TCIAT3703-25G
TCIAT3703-5G
Tetravinyltin ≥98.0%
Tetravinyltin
Formel:
C₈H₁₂Sn Kokpunkt: 163°C Flampunkt: 40°C |
MDL Number:
MFCD00008608 CAS nummer: 1112-56-7 EINECS: 214-193-6 UN: 1992 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...