Acetylthio-PEG4-Alkyne ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
S-3,6,9,12-Tetraoxapentadec-14-yn-1-yl Ethanethioate, Ethanethioic acid S-3,6,9,12-Tetraoxapentadec-14-yn-1-yl ester
T3698-250MG
T3698-50MG
TCIAT3698-250MGEA
8450
SEK
TCIAT3698-250MG
TCIAT3698-50MG
Acetylthio-PEG4-Alkyne ≥98.0%
Acetylthio-PEG4-Alkyne
Formel:
C₁₃H₂₂O₅S MW: 290,37 g/mol Storage Temperature: Frys |
CAS nummer:
1422540-88-2 |
Specifikation testresultat
Appearance | Yellow to Amber to Dark red clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...