Tributylmethylammonium iodide ≥98.0%

Leverantör: TCI
Varning

T3692-5G T3692-25G
TCIAT3692-5GEA 895 SEK
TCIAT3692-5G TCIAT3692-25G
Tributylmethylammonium iodide ≥98.0%
Tributylmethylammonium iodide
Formel: C₁₃H₃₀IN
MDL Number: MFCD00031623
CAS nummer: 3085-79-8
EINECS: 221-402-4

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR