Titanium(IV) 2-ethylhexyloxide
Leverantör: TCI
Varning
Synonyms: Tetrakis(2-ethylhexyl) Orthotitanate, Titanium(IV) 2-Ethylhexyloxide, Tetrakis(2-ethylhexyl)orthotitanate(IV)
T3682-100G T3682-500G
TCIAT3682-100GEA 230 SEK
TCIAT3682-100G TCIAT3682-500G
Titanium(IV) 2-ethylhexyloxide
Titanium(IV) 2-ethylhexyloxide
Formel: C₃₂H₆₈O₄Ti
MW: 564.79 g/mol
Flampunkt: 53°C
MDL Number: MFCD00015264
CAS nummer: 1070-10-6
EINECS: 213-969-1
UN: 1993
ADR: 3 III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Ignition residue 13.4 to 14.6 %