1,2,4-Tribromo-5-iodobenzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,2,4-Tribromo-5-iodobenzene
T3669-1G
T3669-5G
TCIAT3669-1GEA
1475
SEK
TCIAT3669-1G
TCIAT3669-5G
1,2,4-Tribromo-5-iodobenzene ≥98.0%
1,2,4-Tribromo-5-iodobenzene
Formel:
C₆H₂Br₃I MW: 440,70 g/mol |
MDL Number:
MFCD18400058 CAS nummer: 366496-32-4 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...