N,N,N',N'-Tetraphenyl[1,1':4',1'':4'',1'''-quaterphenyl]-4,4'''-diamine ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE

Synonyms: N4,N4,N4''',N4'''-Tetraphenyl-[1,1':4',1'':4'',1'''-quaterphenyl]-4,4'''-diamine

T3657-1G T3657-200MG
TCIAT3657-1GEA 3675 SEK
TCIAT3657-1G TCIAT3657-200MG
N,N,N',N'-Tetraphenyl[1,1':4',1'':4'',1'''-quaterphenyl]-4,4'''-diamine ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetraphenyl[1,1':4',1'':4'',1'''-quaterphenyl]-4,4'''-diamine
Formel: C₄₈H₃₆N₂
MW: 640,83 g/mol
MDL Number: MFCD09833572
CAS nummer: 145898-89-1

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR