Thifluzamide ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2′,6′-Dibromo-2-methyl-4′-trifluoromethoxy-4-trifluoromethyl-1,3-thiazole-5-carboxanilide
T3648-250MG
T3648-1G
TCIAT3648-250MGEA
1625
SEK
TCIAT3648-250MG
TCIAT3648-1G
Thifluzamide ≥98.0%
Thifluzamide
Formel:
C₁₃H₆Br₂F₆N₂O₂S MW: 528,1 g/mol |
MDL Number:
MFCD03428071 CAS nummer: 130000-40-7 UN: 3077 ADR: 9,III |
Specifikation testresultat
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 175.0 to 179.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...