Tebuthiuron ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-[5-(tert-Butyl)-1,3,4-thiadiazol-2-yl]-1,3-dimethylurea
T3647-5G
T3647-25G
TCIAT3647-5GEA
1525
SEK
TCIAT3647-5G
TCIAT3647-25G
Tebuthiuron ≥98.0%
Tebuthiuron
Formel:
C₉H₁₆N₄OS MW: 228,31 g/mol |
MDL Number:
MFCD00078732 CAS nummer: 34014-18-1 EINECS: 251-793-7 UN: 3077 ADR: 9,III Merck Index: 14,09095 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 161.0 to 165.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...