2,2,2-Trifluoro-N-phenylacetamide ≥98.0%
Leverantör: TCI EUROPE
T3645-1G
T3645-5G
TCIAT3645-1GEA
630
SEK
TCIAT3645-1G
TCIAT3645-5G
2,2,2-Trifluoro-N-phenylacetamide ≥98.0%
2,2,2-Trifluoro-N-phenylacetamide
Formel:
C₈H₆F₃NO MW: 189,1346 g/mol |
MDL Number:
MFCD00196027 CAS nummer: 404-24-0 EINECS: 206-965-6 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...