N,N,N',N'-Tetrakis(4-methoxyphenyl)-9H-carbazole-3,6-diamine ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI
Synonyms: 3,6-Bis[N,N-bis(4-methoxyphenyl)amino]-9H-carbazole
T3642-1G T3642-5G
TCIAT3642-1GEA 6400 SEK
TCIAT3642-1G TCIAT3642-5G
N,N,N',N'-Tetrakis(4-methoxyphenyl)-9H-carbazole-3,6-diamine ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetrakis(4-methoxyphenyl)-9H-carbazole-3,6-diamine
Formel: C₄₀H₃₅N₃O₄
MW: 621.74 g/mol
CAS nummer: 1630723-98-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7da9df358b1a1fe49e4ba46f5f330266178c92f0&vid=7da9df358b1a1fe49e4ba46f5f330266178c92f0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %