Thiophen-2-carbothioamide ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Thiophene-2-carbothioamide
T3604-5G
T3604-25G
TCIAT3604-5GEA
4250
SEK
TCIAT3604-5G
TCIAT3604-25G
Thiophen-2-carbothioamide ≥98.0%
Thiophen-2-carbothioamide
Formel:
C₅H₅NS₂ MW: 143,23 g/mol |
MDL Number:
MFCD00052890 CAS nummer: 20300-02-1 |
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Melting point | 101.0 to 105.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...