Trifluoromethanesulphonyl bromide ≥95.0%
Leverantör: TCI
|
Fara
|
T3592-1G
T3592-5G
TCIAT3592-1GEA
855
SEK
TCIAT3592-1G
TCIAT3592-5G
Trifluoromethanesulphonyl bromide ≥95.0%
Trifluoromethanesulphonyl bromide
Formel:
CBrF₃O₂S Storage Temperature: Kylskåp |
CAS nummer:
15458-53-4 UN: 3265 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...