Tridecan-7-amine ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Hexylheptylamine, 7-Aminotridecane

T3591-1ML T3591-5ML
TCIAT3591-1MLEA 2025 SEK
TCIAT3591-1ML TCIAT3591-5ML
Tridecan-7-amine ≥97.0%
Tridecan-7-amine
Formel: C₁₃H₂₉N
MW: 199,38 g/mol
Kokpunkt: 102°C
MDL Number: MFCD20690568
CAS nummer: 22513-16-2
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62131fe12af51cc9affc1911d0e663be4315bbee&vid=62131fe12af51cc9affc1911d0e663be4315bbee&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR