Triethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane ≥98.0%
Leverantör: TCI EUROPE
Synonyms: (2-(7-Oxabicyclo[4.1.0]heptan-3-yl)ethyl)triethoxysilane, 2-(3,4-Epoxycyclohexyl)ethyltriethoxysilane
T3585-5ML T3585-25ML
TCIAT3585-5MLEA 215 SEK
TCIAT3585-5ML TCIAT3585-25ML
Triethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane ≥98.0%
Triethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane
Formel: C₁₄H₂₈O₄Si
MW: 288,46 g/mol
Kokpunkt: 130°C
MDL Number: MFCD00069159
CAS nummer: 10217-34-2
EINECS: 425-050-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=088dcc365ee3655f0855784d19df3967bc8c8881&vid=088dcc365ee3655f0855784d19df3967bc8c8881&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.0050 to 1.0150