TOTT ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-(((Dimethylamino)(dimethyliminio)methyl)thio)pyridine 1-oxide tetrafluoroborate, N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiouronium Tetrafluoroborate, TOTT

T3569-5G T3569-25G
TCIAT3569-5GEA 1300 SEK
TCIAT3569-5G TCIAT3569-25G
TOTT ≥98.0%
TOTT
Formel: C₁₀H₁₆BF₄N₃OS
MW: 313,122 g/mol
Storage Temperature: Frys
MDL Number: MFCD05664718
CAS nummer: 255825-38-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0298db233a18c39f0c3bb2bd29d5ab10d1e6fc4&vid=a0298db233a18c39f0c3bb2bd29d5ab10d1e6fc4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR