1H,1H,2H,2H-Perfluorooctyltrimethoxysilane ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Trimethoxy(1H,1H,2H,2H-perfluoro-n-octyl)silane, Trimethoxy(1H,1H,2H,2H-tridecafluoro-n-octyl)silane, Trimethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
T3560-5G T3560-25G
TCIAT3560-5GEA 1125 SEK
TCIAT3560-5G TCIAT3560-25G
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane ≥97.0%
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane
Formel: C₁₁H₁₃F₁₃O₃Si
MW: 468,2800 g/mol
Kokpunkt: 71°C
MDL Number: MFCD00153261
CAS nummer: 85857-16-5
EINECS: 288-657-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57146b72527b2bf2e4d438cf10dc4af695c9c97b&vid=57146b72527b2bf2e4d438cf10dc4af695c9c97b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %