Triallyl(methyl)silane ≥95.0%

Leverantör: TCI
Varning

T3549-1ML T3549-5ML
TCIAT3549-1MLEA 565 SEK
TCIAT3549-1ML TCIAT3549-5ML
Triallyl(methyl)silane ≥95.0%
Triallyl(methyl)silane
Formel: C₁₀H₁₈Si
Kokpunkt: 180°C
MDL Number: MFCD00800604
CAS nummer: 1112-91-0
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02a95031ecf4e7eb0c95b55594819f445fdbcc71&vid=bc153295b0f36a19afa0a58f4aa74fdb08cc5771&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR