2-Aminophenylboronic acid pinacol ester ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester
T3543-1G
T3543-5G
TCIAT3543-1GEA
575
SEK
TCIAT3543-1G
TCIAT3543-5G
2-Aminophenylboronic acid pinacol ester ≥98.0%
2-Aminophenylboronic acid pinacol ester
Formel:
C₁₂H₁₈BNO₂ MW: 219.09 g/mol Kokpunkt: 125°C |
MDL Number:
MFCD02179448 CAS nummer: 191171-55-8 |
|
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...