Phenethyltrimethoxysilane ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Trimethoxy(phenethyl)silane, Trimethoxy(2-phenylethyl)silane [contains ca. 25% Trimethoxy(1-phenylethyl)silane], Trimethoxy(2-phenylethyl)silane
T3542-25G
T3542-100G
TCIAT3542-25GEA
1125
SEK
TCIAT3542-25G
TCIAT3542-100G
Phenethyltrimethoxysilane ≥95.0%
Phenethyltrimethoxysilane
Formel:
C₁₁H₁₈O₃Si MW: 226,347 g/mol Kokpunkt: 122°C Flampunkt: 109°C |
MDL Number:
MFCD00053852 CAS nummer: 49539-88-0 EINECS: 256-363-2 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Red clear liquid |
Purity (GC) | min. 95.0 % (total of isomer) |
Specific gravity (20/20) | 1.0390 to 1.0440 |
Refractive index n20/D | 1.4750 to 1.4800 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...