1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI

Synonyms: 2,2',2''-(1,3,5-Benzenetriyl)tris(1-phenyl-1H-benzimidazole), TPBi, 1,3,5-Tris(1-phenyl-1H-benzo[d]imidazol-2-yl)benzene

T3537-200MG T3537-1G
TCIAT3537-200MGEA 710 SEK
TCIAT3537-200MG TCIAT3537-1G
1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene ≥98.0% (by HPLC, titration analysis)
1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene
Formel: C₄₅H₃₀N₆
MW: 654.78 g/mol
MDL Number: MFCD08276433
CAS nummer: 192198-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19080411087580dac8ee91c6a5b37bd0833cd4ee&vid=19080411087580dac8ee91c6a5b37bd0833cd4ee&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR