1,3,5-Tris(trimethylsilyl)benzene ≥95.0%
Leverantör: TCI EUROPE
T3526-1G
T3526-5G
TCIAT3526-1GEA
1175
SEK
TCIAT3526-1G
TCIAT3526-5G
1,3,5-Tris(trimethylsilyl)benzene ≥95.0%
1,3,5-Tris(trimethylsilyl)benzene
Formel:
C₁₅H₃₀Si₃ MW: 294,66 g/mol Kokpunkt: 244°C |
MDL Number:
MFCD00093247 CAS nummer: 5624-60-2 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...