5-Cyano-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile, 3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Cyanopyridine-5-boronic acid Pinacol ester, 3-Cyanopyridine-5-boronic acid pinacol ester, 2-(5-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
T3514-200MG
T3514-1G
TCIAT3514-200MGEA
430
SEK
TCIAT3514-200MG
TCIAT3514-1G
5-Cyano-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
5-Cyano-3-pyridineboronic acid pinacol ester
Formel:
C₁₂H₁₅BN₂O₂ MW: 230,07 g/mol |
MDL Number:
MFCD07780755 CAS nummer: 402718-29-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...