Tofisopam ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-(3,4-Dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-2,3-benzodiazepine, 1-(3,4-Dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-benzo[d][1,2]diazepine
T3509-50MG
T3509-250MG
TCIAT3509-50MGEA
950
SEK
TCIAT3509-50MG
TCIAT3509-250MG
Tofisopam ≥98.0%
Tofisopam
Formel:
C₂₂H₂₆N₂O₄ MW: 382,46 g/mol Smältpunkt: 157°C |
MDL Number:
MFCD00823171 CAS nummer: 22345-47-7 EINECS: 244-922-3 UN: 3077 ADR: 9,III Merck Index: 14,09503 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 7.00 to 7.70 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...