Tetraetylammoniumväte ulfat ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Tetraethylammonium bisulphate, Tetraethylammonium hydrogensulphate

T3506-5G T3506-25G
TCIAT3506-5GEA 645 SEK
TCIAT3506-5G TCIAT3506-25G
Tetraetylammoniumväte ulfat ≥98.0%
Tetraetylammoniumväte ulfat
Formel: C₈H₂₁NO₄S
MW: 227,33 g/mol
MDL Number: MFCD00036150
CAS nummer: 16873-13-5
EINECS: 240-899-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR