4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide ≥95.0%

Leverantör: TCI EUROPE

Synonyms: A-205804, 4-[(4-Methylphenyl)thio]thieno[2,3-c]pyridine-2-carboxamide, A 205804

T3498-100MG T3498-25MG
TCIAT3498-100MGEA 5975 SEK
TCIAT3498-100MG TCIAT3498-25MG
4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide ≥95.0%
4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide
Formel: C₁₅H₁₂N₂OS₂
MW: 300,39 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09038566
CAS nummer: 251992-66-2

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 8.80 to 9.80 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR