Tetraethylene glycol dimethacrylate ≥90.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: ((Oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) bis(2-methylacrylate)
Stabilization: med hydrokinon
T3486-25G T3486-100G
TCIAT3486-25GEA 565 SEK
TCIAT3486-25G TCIAT3486-100G
Tetraethylene glycol dimethacrylate ≥90.0% stabilised
Tetraethylene glycol dimethacrylate
Formel: C₁₆H₂₆O₇
MW: 330,38 g/mol
Kokpunkt: 200°C
Flampunkt: 82°C
Storage Temperature: Kylskåp
MDL Number: MFCD00014932
CAS nummer: 109-17-1
EINECS: 203-653-1
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %