4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (2-Methyl-1-propenyl)boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(2-methylprop-1-en-1-yl)-1,3,2-dioxaborolane

T3469-1G T3469-5G
TCIAT3469-1GEA 470 SEK
TCIAT3469-1G TCIAT3469-5G
4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane ≥98.0%
4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane
Formel: C₁₀H₁₉BO₂
MW: 182,07 g/mol
Kokpunkt: 71°C
Flampunkt: 66°C
MDL Number: MFCD09842762
CAS nummer: 126689-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e169540d317e90ebde6077effa7f57201b0a8d63&vid=e169540d317e90ebde6077effa7f57201b0a8d63&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Specific gravity (20/20) 0.8960 to 0.9000

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR