1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate ≥98.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate, Acrylic acid 1H,1H,2H,2H-Tridecafluoro-n-octyl ester
Stabilization: Stabilised with HQ + MEHQ
T3451-5G T3451-25G
TCIAT3451-5GEA 965 SEK
TCIAT3451-5G TCIAT3451-25G
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate ≥98.0% stabilised
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate
Formel: C₁₁H₇F₁₃O₂
MW: 418,154 g/mol
Kokpunkt: 50°C
Flampunkt: 102°C
MDL Number: MFCD00042351
CAS nummer: 17527-29-6
EINECS: 241-527-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d1f1331a5a5b11c8739bb53d520681293a2af3d&vid=9d1f1331a5a5b11c8739bb53d520681293a2af3d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %