[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Mesityl[4-(trifluoromethyl)phenyl]iodonium triflate, [4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium Trifluoromethanesulphonate
T3446-1G
T3446-5G
TCIAT3446-1GEA
1325
SEK
TCIAT3446-1G
TCIAT3446-5G
[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0%
[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate
Formel:
C₁₇H₁₅F₆IO₃S MW: 540,26 g/mol |
MDL Number:
MFCD27978254 CAS nummer: 1232133-62-8 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Melting point | 201.0 to 205.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...