Tri(cyclohexa-2,5-dien-1-yl)silane ≥93.0%

Leverantör: TCI
Varning

T3443-200MG T3443-1G
TCIAT3443-200MGEA 870 SEK
TCIAT3443-200MG TCIAT3443-1G
Tri(cyclohexa-2,5-dien-1-yl)silane ≥93.0%
Tri(cyclohexa-2,5-dien-1-yl)silane
Formel: C₁₈H₂₂Si
CAS nummer: 1807776-35-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c313dc238fa1e8726e38ac7211d95d0392b05cea&vid=c313dc238fa1e8726e38ac7211d95d0392b05cea&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 93.0 %
Melting point 63.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR