2,3,6-Trichloropyridine ≥97.0%
Leverantör: TCI EUROPE
Varning
T3428-25G T3428-5G
TCIAT3428-25GEA 2550 SEK
TCIAT3428-25G TCIAT3428-5G
2,3,6-Trichloropyridine ≥97.0%
2,3,6-Trichloropyridine
Formel: C₅H₂Cl₃N
MW: 182,43 g/mol
Kokpunkt: 234°C
MDL Number: MFCD00463516
CAS nummer: 6515-09-9
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 64.0 to 68.0 deg-C