2',3,4-Trifluoro-4''-propyl-1,1':4',1''-terphenyl ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2',3,4-Trifluoro-4''-propyl-p-terphenyl

T3419-1G T3419-5G
TCIAT3419-1GEA 910 SEK
TCIAT3419-1G TCIAT3419-5G
2',3,4-Trifluoro-4''-propyl-1,1':4',1''-terphenyl ≥98.0%
2',3,4-Trifluoro-4''-propyl-1,1':4',1''-terphenyl
Formel: C₂₁H₁₇F₃
MW: 326,36 g/mol
MDL Number: MFCD20488039
CAS nummer: 248936-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1e8a466c116394b7e205155876d509e5df1f7a1&vid=d1e8a466c116394b7e205155876d509e5df1f7a1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 71.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR