1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1,3,5-Trimethylpyrazole-4-boronic acid Pinacol ester, 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(1,3,5-Trimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
T3409-200MG
T3409-1G
TCIAT3409-200MGEA
510
SEK
TCIAT3409-200MG
TCIAT3409-1G
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester
Formel:
C₁₂H₂₁BN₂O₂ MW: 236.12 g/mol |
MDL Number:
MFCD06659062 CAS nummer: 844891-04-9 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...