4-([2,2':6',2''-Terpyridin]-4'-yl)benzoic acid ≥98.0%

Leverantör: TCI
Varning

Synonyms: 4'-(4-Carboxyphenyl)-2,2':6',2''-terpyridine

T3405-200MG T3405-1G
TCIAT3405-200MGEA 895 SEK
TCIAT3405-200MG TCIAT3405-1G
4-([2,2':6',2''-Terpyridin]-4'-yl)benzoic acid ≥98.0%
4-([2,2':6',2''-Terpyridin]-4'-yl)benzoic acid
Formel: C₂₂H₁₅N₃O₂
MDL Number: MFCD22056799
CAS nummer: 158014-74-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a729e92f13602a4bca3ecee1fcddd006ca9d17fc&vid=a729e92f13602a4bca3ecee1fcddd006ca9d17fc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR