1H,1H-Tricosafluoro-1-dodecanol ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Tricosafluor-1-dodecanol, 1H,1H-Perfluoro-1-dodecanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Tricosafluoro-1-dodecanol
T3381-1G
T3381-5G
TCIAT3381-1GEA
310
SEK
TCIAT3381-1G
TCIAT3381-5G
1H,1H-Tricosafluoro-1-dodecanol ≥98.0%
1H,1H-Tricosafluoro-1-dodecanol
Formel:
C₁₂H₃F₂₃O MW: 600,12 g/mol Kokpunkt: 224°C |
MDL Number:
MFCD00153235 CAS nummer: 423-65-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...